Learn More
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes in Tube Packs
For biopsy tissue samples, these cassettes have a back-edge lid hinge, loose-packed lids, and are compatible with all Epredia PrintMate™ Cassette Printers
Brand:  Epredia™ B851750LG
 View more versions of this product
Product Code. 16314156
  • £261.00 / Pack of 750
Explore more special offers
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Small Squares
750/Cs.
Yes
For Use With PrintMate™ Printers
Yes
Separate Lid, Back Hinge
Light Green
TissueLoc Biopsy Cassette