Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes in Tube Packs
Slotted Cassettes are ideal for use with routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in twelve color varieties.
Brand:  Epredia™ B851720LA
78.00 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 16334046
  • £109.00 / Pack of 750
Explore more special offers
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Slots
750/Cs.
Yes
For Use With PrintMate™ Printers
Yes
Separate Lid, Back Hinge
Lavender
TissueLoc™ Slotted Cassette