missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ TissueLoc™ Tissue Cassettes, White

Product Code. p-6024290 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Peach
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Pack of 500
This item is not returnable. View return policy

Product Code. 12068838

Brand: Epredia™ 58958

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Epredia™ TissueLoc™ Tissue Cassettes.

  • Three-sided writing surface ensures positive identification from grossing to filing
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Eleven colors allow for easy specimen coding
TRUSTED_SUSTAINABILITY

Specifications

Pore Style Slotted Pore
Closure Type Hinged
Color Pink
Product Type Tissue Cassette
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.