missing translation for 'onlineSavingsMsg'
Learn More

Titanium aluminide, 99.5% (metals basis)

Product Code. p-7076345
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11368117

Brand: Thermo Scientific Alfa Aesar 022895.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products. | CAS: 12004-78-3 | Al3Ti | 128.812 g/mol

Titanium aluminide is used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-78-3
Molecular Formula Al3Ti
Molecular Weight (g/mol) 128.812
MDL Number MFCD00054094
InChI Key OQPDWFJSZHWILH-UHFFFAOYSA-N
Synonym titanium aluminum, aluminum, compd. with titanium 3:1, aluminum; titanium, aluminum, compound with titanium 3:1
PubChem CID 6336852
IUPAC Name aluminum;titanium
SMILES [Al].[Al].[Al].[Ti]

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN3178
Solubility Information Insoluble in water.
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Titanium aluminide
compliance-icons compliance-icons
Product Identifier
  • Titanium aluminide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.