missing translation for 'onlineSavingsMsg'
Learn More

Titanium boride, Thermo Scientific Chemicals

Product Code. 11371139
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Titanium boride is used in seals wear parts and cutting tools. It is used to make ballistic armour due to its high hardness. It is used to constitute composite materials in which the presence of the material serves to increase strength and fracture toughness of the matrix.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-63-5
Molecular Formula B2Ti
Molecular Weight (g/mol) 69.487
MDL Number MFCD00049595
InChI Key QYEXBYZXHDUPRC-UHFFFAOYSA-N
Synonym titanium boride,
PubChem CID 131664302
IUPAC Name boron;titanium
SMILES [B].[B].[Ti]

Specifications

Melting Point 3225°C
Physical Form Powder
Quantity 1 kg
Solubility Information Soluble in hydrogen peroxide and sulfuric acid.
Formula Weight 69.5
Density 4.38
Chemical Name or Material Titanium Boride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'