missing translation for 'onlineSavingsMsg'
Learn More

Titanium boride, Thermo Scientific Chemicals

£36.48 - £296.48

Chemical Identifiers

CAS 12045-63-5
Molecular Formula B2Ti
Molecular Weight (g/mol) 69.487
MDL Number MFCD00049595
InChI Key QYEXBYZXHDUPRC-UHFFFAOYSA-N
Synonym titanium boride,
PubChem CID 131664302
IUPAC Name boron;titanium
SMILES [B].[B].[Ti]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381139
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011364.18
50 g
£36.48
50g
Please to purchase this item. Need a web account? Register with us today!
11391139
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011364.30
250 g
£103.52
250g
Please to purchase this item. Need a web account? Register with us today!
11371139
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011364.A1
1 kg
£296.48
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Titanium boride is used in seals wear parts and cutting tools. It is used to make ballistic armour due to its high hardness. It is used to constitute composite materials in which the presence of the material serves to increase strength and fracture toughness of the matrix.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12045-63-5
69.487
QYEXBYZXHDUPRC-UHFFFAOYSA-N
131664302
[B].[B].[Ti]
B2Ti
MFCD00049595
titanium boride,
boron;titanium

Specifications

3225°C
Powder
MFCD00049595
Soluble in hydrogen peroxide and sulfuric acid.
[B].[B].[Ti]
69.487
69.5
Titanium Boride
12045-63-5
B2Ti
titanium boride,
QYEXBYZXHDUPRC-UHFFFAOYSA-N
boron;titanium
131664302
4.38
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-961-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only