missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium carbide, nanopowder, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Titanium carbide is used in the manufacture of wear-resistant tools, cutting tools, As a coating for abrasive steel bearings, In the form of nano titanium carbide ceramic in optics applications, Enhance the conductivity of materials and as a nucleating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 4820°C |
Melting Point | ∽3140°C |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Soluble in nitric acid and aqua regia. Insoluble in water. |
Formula Weight | 59.91 |
Density | 4.93 |
Chemical Name or Material | Titanium carbide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'