missing translation for 'onlineSavingsMsg'
Learn More
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 15425945

Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 15425945
100 x 2000 mm
Click to view available options
Quantity:
100 x 100 mm
100 x 500 mm
100 x 2000 mm
This item is not returnable. View return policy

Product Code. 15425945

Brand: Thermo Scientific Alfa Aesar 010385.HY

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 11.3g/100x100mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Especificaciones

Color Silver to Gray
Appearance Silver Gray foil
Assay Percent Range 99.5% (Metals basis)
Quantity 100 x 2000 mm
Odor Odorless
Form Foil
Chemical Name or Material Titanium, Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW10K009 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number010385.GH,010385.HY,010385.HP
Lot NumberN14K004 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number010385.GH,010385.HY,010385.HP
Lot NumberN14K005 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number010385.GH,010385.HY,010385.HP
Lot NumberS14H049 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog Number010385.GH,010385.HY,010385.HP
Lot NumberM31I039 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010385.GH,010385.HY,010385.HP

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado