missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) hydride, Ti 94.0% min.

Product Code. 11369399
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
This item is not returnable. View return policy

Product Code. 11369399

Brand: Thermo Scientific Alfa Aesar 089183.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7704-98-5 | H2Ti

For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H{2}Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-98-5
Molecular Formula H2Ti
MDL Number MFCD00151301

Specifications

Melting Point 450°C (decomposition)
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%, metal basis
Solubility Information Soluble in water.
Formula Weight 49.92
Sensitivity Air and moisture sensitive
Density 3.9
Chemical Name or Material Titanium(II) Hydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.