missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) hydride, 99% (metals basis)

Product Code. 11307597
Click to view available options
Quantity:
5 g
100 g
500 g
2 x 1 kg
Unit Size:
100g
2kg
500g
5g
This item is not returnable. View return policy

Product Code. 11307597

Brand: Thermo Scientific Alfa Aesar 012857.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Widley used as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. | CAS: 7704-98-5 | H2Ti

Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-98-5
Molecular Formula H2Ti
MDL Number MFCD00151301

Specifications

Melting Point 450°C (decomposition)
Physical Form Powder
Quantity 100 g
Linear Formula TiH2
UN Number UN1871
Solubility Information Soluble in water.
Formula Weight 49.92
Percent Purity 99%
Packaging Plastic bottle
Sensitivity Air and moisture sensitive
Specific Gravity 3.9
Density 3.9
Chemical Name or Material Titanium(II) hydride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.