Learn More
Titanium(II) oxide pellets, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Used for vacuum deposition. Also applications include sunscreens and UV-blocking in cosmetics, varnishes, wood preservatives, textile fibers, and packaging films. Also used as paint pigment, opacifying agent, welding rod fluxes, optical coatings, as a catalyst, ceramic finish coat, plastics, elastomers, coated fabrics, printing inks, roofing granules, glass, and in glazes. Also identified as Titanium monoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | >3000°C |
Melting Point | 1700°C |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 63.9 |
Percent Purity | 99.9% |
Odor | Odorless |
Density | 4.95 g/mL |
Assay | (metals basis) |
Chemical Name or Material | Titanium (II) oxide pellets, 8 to 9mm(0.32 to 0.36 in.) dia. x 6 to 7mm(0.24 to 0.28 in.) thick |
RUO – Research Use Only