Learn More
Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 429151000
70.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 29
Additional Details : CAS Number : 7705-07-9 Weight : 0.12200kg Transport : UN number : 3264 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Titanium(III) chloride | |
Purple | |
100 mL | |
TiCl3 | |
15, 9637 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether | |
[Cl-].[Cl-].[Cl-].[Ti+3] | |
154.22 | |
∼20% (TiCl3) (exact strength stated on certificate of analysis) | |
1.2000g/mL |
7705-07-9,7647-01-0,7732-18-5 | |
Liquid | |
Cl3Ti | |
MFCD00011266 | |
Titanous chloride | |
YONPGGFAJWQGJC-UHFFFAOYSA-K | |
titanium(3+) trichloride | |
154.26 | |
AcroSeal™ Glass Bottle |
Safety and Handling
- Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
- MIXTURE LIST-Contains : Titanium trichloride, Hydrochloric acid,Phosphoric acid
RUO – Research Use Only