missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid

Product Code. p-3593535 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Plastic bottle
Unit Size:
1 L
250mL
25mL
This item is not returnable. View return policy

Product Code. 10247043

Brand: Thermo Scientific Acros 395620250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7705-07-9 | Cl3Ti | 154.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Titanium(III) chloride
CAS 7705-07-9, 7647-01-0, 7732-18-5
Color Purple
Physical Form Liquid
Quantity 25 mL
Molecular Formula Cl3Ti
Linear Formula TiCl3
MDL Number MFCD00011266
Merck Index 15,9637
Synonym Titanous chloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether
InChI Key YONPGGFAJWQGJC-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Ti+3]
IUPAC Name titanium(3+) trichloride
Molecular Weight (g/mol) 154.22
Formula Weight 154.26
Percent Purity ∼20% (TiCl3) (exact strength stated on certificate of analysis)
Packaging Plastic bottle
Density 1.2000g/mL
Show More Show Less
compliance-icons
Product Identifier
  • Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
  • MIXTURE LIST-Contains : Titanium trichloride, Hydrochloric acid,Phosphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.