missing translation for 'onlineSavingsMsg'
Learn More
Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%, Thermo Scientific Chemicals Product Code.: 11304257

Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%, Thermo Scientific Chemicals

Product Code. 11304257
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11304257

Brand: Thermo Scientific Alfa Aesar 036729.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a catalyst in the production of polypropylene.

Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5% is used as a catalyst in the production of polypropylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-13-3
Molecular Formula AlCl12Ti3
Molecular Weight (g/mol) 595.98
MDL Number MFCD00145412
InChI Key KSKPGAGLAZXNEO-UHFFFAOYSA-H
IUPAC Name titanium(4+) aluminium(3+) heptachloride
SMILES Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl

Specifications

Physical Form Powder
Quantity 100 g
Assay Percent Range 76.0 to 78.5%
Synonym Titanium aluminum chloride
Solubility Information Insoluble in water.
Formula Weight 596.15
Sensitivity Air and moisture sensitive
Chemical Name or Material Titanium(III) Chloride-Aluminum(III) Chloride
compliance-icons
Product Identifier
  • Titanium(III) chloride-Aluminum(III) chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.