Learn More
Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals
£30.80 - £133.00
Chemical Identifiers
CAS | 18039-90-2 |
---|---|
Molecular Formula | C12H24Cl3O3Ti |
Molecular Weight (g/mol) | 370.56 |
MDL Number | MFCD01631276 |
InChI Key | MIDYWIPTCYRMQF-UHFFFAOYSA-K |
Synonym | oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3 |
PubChem CID | 6097232 |
IUPAC Name | oxolane;trichlorotitanium |
SMILES | C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10525983
|
Thermo Scientific Acros
375160010 |
1 g | Glass bottle |
£30.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10598663
|
Thermo Scientific Acros
375160100 |
10 g | Glass Bottle |
£133.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
18039-90-2 | |
370.56 | |
MIDYWIPTCYRMQF-UHFFFAOYSA-K | |
6097232 | |
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl |
C12H24Cl3O3Ti | |
MFCD01631276 | |
oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3 | |
oxolane;trichlorotitanium |
Specifications
>250°C | |
Blue to Blue-Green | |
96% min. (Argentometry) | |
MFCD01631276 | |
Solubility in water: reacts violently | |
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl | |
370.56 | |
370.56 | |
Authentic |
18039-90-2 | |
Crystals or Powder | |
C12H24Cl3O3Ti | |
oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3 | |
MIDYWIPTCYRMQF-UHFFFAOYSA-K | |
oxolane;trichlorotitanium | |
6097232 | |
97% | |
Titanium(III) chloride tetrahydrofuran complex |
Safety and Handling
GHS H Statement:
Flammable solid.
Causes serious eye damage.
Reacts violently with water.
May form explosive peroxides.
Contact with water liberates toxic gas.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Titanium(III) chloride tetrahydrofuran complex
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only