missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) bromide, 98%

Product Code. 10085143 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10085143

Brand: Thermo Scientific Acros 391240250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-68-6 | TiBr4 | 367.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-68-6
Molecular Formula TiBr4
Molecular Weight (g/mol) 367.5
MDL Number MFCD00011265

Specifications

Boiling Point 230.0°C
Melting Point 39.0°C
Quantity 25 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula TiBr4
Merck Index 15,9633
Solubility Information Solubility in water: reacts. Other solubilities: soluble in absolute ether and absolute alcohol
Formula Weight 367.5
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Titanium(IV) bromide
Show More Show Less
compliance-icons
Product Identifier
  • Titanium(IV) bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.