missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals Product Code.: 15417915

Titanium(IV) chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals

Product Code. 15417915
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 15417915

Brand: Thermo Scientific Alfa Aesar H31830.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-45-0
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4Show MoreShow Less
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Quantity 100 mL
Linear Formula TiCl4
UN Number UN2922
Solubility Information Reacts with water.
Formula Weight 189.69
Concentration or Composition (by Analyte or Components) 1M soln. in dichloromethane
Sensitivity Moisture sensitive; May form a precipitate
Density 1.365
Chemical Name or Material Titanium(IV) chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247652 Certificate TypeCertificate of Analysis Date18/08/2023 Catalog NumberH31830.AE,H31830.AP
Lot Number10239990 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH31830.AE,H31830.AP
Lot Number10170914 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31830.AE,H31830.AP
Lot Number10222303 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31830.AE,H31830.AP
Lot Number10202020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31830.AE,H31830.AP
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) chloride, 1M solution in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dichloromethane, Titanium tetrachloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.