Learn More
Titanium(IV) chloride, 99.0% min, typically 99.6%, Thermo Scientific Chemicals
Description
In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screenTitanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 136°C to 137°C |
Melting Point | 25°C |
Color | Yellow |
Physical Form | Liquid |
Quantity | 100 mL |
Assay Percent Range | 99.0% min., typically 99.6% |
Solubility Information | Soluble in dilute hydrochloric acid,alcohol and alkalies.Miscible with ethanol,hydrochloric acid and organic solvents. |
Formula Weight | 189.69 |
Refractive Index | 1.61 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only