missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) chloride, 99.0% min, typically 99.6%, Thermo Scientific Chemicals

Product Code. 15411077
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use in Manufacture of Titanium Pigments and Metal, As a Catalyst in organic Syntheses, in Iridescent Glass

In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screenTitanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-45-0
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Boiling Point 136°C to 137°C
Melting Point °C to 25°C
Color Yellow
Physical Form Liquid
Quantity 1 L
Assay Percent Range 99.0% min., typically 99.6%
Solubility Information Soluble in dilute hydrochloric acid,alcohol and alkalies.Miscible with ethanol,hydrochloric acid and organic solvents.
Formula Weight 189.69
Refractive Index 1.61
Sensitivity Moisture sensitive
Appearance Colorless to yellow liquid
Density 1.726
Chemical Name or Material Titanium(IV) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'