missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) chloride, 99.9%, Thermo Scientific Chemicals Product Code.: 10184210

Titanium(IV) chloride, 99.9%, Thermo Scientific Chemicals

Product Code. 10184210
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10184210

Brand: Thermo Scientific Acros 197231000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-45-0
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4Show MoreShow Less
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Boiling Point 136.5°C
Melting Point -24.0°C
Color Yellow
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 99.85% min
Molecular Formula Cl4Ti
Linear Formula TiCl4
Fieser 01,1169; 02,414; 03,291; 04,507; 05,671; 06,590; 07,370; 08,483; 09,468; 10,401; 11,337; 12,94; 13,6; 14,111; 15,317; 16,332; 17,344
Merck Index 15, 9634
Solubility Information Solubility in water: reacts forming hcl. Other solubilities: miscible with cold cond. hcl, soluble in alcohol (reacts, forming hcl), miscible with aliphatic hydrocarbons and, chlorinated aliphatic hydrocarbons
Molecular Weight (g/mol) 189.71
Formula Weight 189.71
Percent Purity 99.9%
Packaging Glass bottle
Specific Gravity 1.72
Density 1.7200g/mL
Chemical Name or Material Titanium(IV) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465837 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number197230010,197231000,197232500
Lot NumberA0464053 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog Number197230010,197231000,197232500
Lot NumberA0445293 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number197230010,197231000,197232500
Lot NumberA0456289 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number197230010,197231000,197232500
Lot NumberA0450895 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number197230010,197231000,197232500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) chloride, 99.9%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.