Learn More
Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals
Description
Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 150°C to 152°C (10mmHg) |
Melting Point | -40°C |
Color | Yellow |
Physical Form | Liquid |
Quantity | 50 g |
Linear Formula | Ti(OCH2CH3)4 |
UN Number | UN2924 |
Flash Point | 28°C (84°F) |
Solubility Information | Soluble in organic solvents; Decomposes in water. |
Formula Weight | 228.15 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
R17K003 | Certificate of Analysis | 11/06/2024 | 077142.30,077142.06,077142.18 |
N23K003 | Certificate of Analysis | 15/03/2024 | 077142.30,077142.06,077142.18 |
N29K014 | Certificate of Analysis | 15/03/2024 | 077142.30,077142.06,077142.18 |
N23K004 | Certificate of Analysis | 15/03/2024 | 077142.30,077142.06,077142.18 |
62200053 | Certificate of Analysis | 20/06/2022 | 077142.30,077142.06,077142.18 |
Safety and Handling


- Titanium(IV) ethoxide, TiO2 33% min
- Warning
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
- MIXTURE LIST-Contains : Titanium(IV) ethoxide, Titanium(IV) isopropoxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.