missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals

Catalog Number 15492957
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy
3087-36-3
C8H20O4Ti
228.111
MFCD00009071
JMXKSZRRTHPKDL-UHFFFAOYSA-N
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
76524
ethanolate;titanium(4+)
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
This item is not returnable. View return policy
3087-36-3
C8H20O4Ti
228.111
MFCD00009071
JMXKSZRRTHPKDL-UHFFFAOYSA-N
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
76524
ethanolate;titanium(4+)
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 150°C to 152°C (10mmHg)
Melting Point -40°C
Color Yellow
Physical Form Liquid
Quantity 5 g
Linear Formula Ti(OCH2CH3)4
UN Number UN2924
Flash Point 28°C (84°F)
Solubility Information Soluble in organic solvents; Decomposes in water.
Formula Weight 228.15
Concentration or Composition (by Analyte or Components) TiO2 33% min
Odor Alcohol-like
Refractive Index 1.5043
Sensitivity Moisture sensitive
Density 1.088 g/mL
Chemical Name or Material Titanium (IV) ethoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) ethoxide, TiO2 33% min
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : Titanium(IV) ethoxide, Titanium(IV) isopropoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'