missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) isobutoxide

Product Code. 15413007
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 15413007

Brand: Thermo Scientific Alfa Aesar 089455.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7425-80-1 | C16H36O4Si | 320.545 g/mol

Titanium(IV) isobutoxide general application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7425-80-1
MDL Number MFCD00015045
Synonym tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester
PubChem CID 69626
IUPAC Name tetrakis(2-methylpropyl) silicate

Specifications

Boiling Point 296°C
Color Yellow
Physical Form Melt
Quantity 50 g
Molecular Formula C16H36O4Si
Linear Formula Ti[(CH3)2CHCH2O]4
Flash Point 77°C (171°F)
Solubility Information Reacts with water.
InChI Key QKCMIPQMYBKEPP-UHFFFAOYSA-N
SMILES CC(C)CO[Si](OCC(C)C)(OCC(C)C)OCC(C)C
Molecular Weight (g/mol) 320.545
Formula Weight 340.34
Refractive Index 1.495
Sensitivity Moisture sensitive
Density 1.02 g/mL
Chemical Name or Material Titanium (IV) isobutoxide
Show More Show Less
compliance-icons
Product Identifier
  • Titanium(IV) isobutoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Titanium(IV) isobutoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.