Learn More
Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids
£26.43 - £229.14
Chemical Identifiers
CAS | 546-68-9 |
---|---|
Molecular Formula | C12H28O4Ti |
Molecular Weight (g/mol) | 284.219 |
MDL Number | MFCD00008871 |
InChI Key | VXUYXOFXAQZZMF-UHFFFAOYSA-N |
Synonym | titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate |
PubChem CID | 11026 |
IUPAC Name | propan-2-olate;titanium(4+) |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15422957
|
Thermo Scientific Alfa Aesar
077115.09 |
10 g |
£26.43
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432957
|
Thermo Scientific Alfa Aesar
077115.22 |
100 g |
£35.88
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15442957
|
Thermo Scientific Alfa Aesar
077115.36 |
500 g |
£88.04
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452957
|
Thermo Scientific Alfa Aesar
077115.57 |
4 x 500 g |
£229.14
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids.Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
546-68-9 | |
284.219 | |
VXUYXOFXAQZZMF-UHFFFAOYSA-N | |
11026 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4] |
C12H28O4Ti | |
MFCD00008871 | |
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate | |
propan-2-olate;titanium(4+) |
Specifications
232°C | |
546-68-9 | |
Liquid | |
Ti[OCH(CH3)2]4 | |
UN2413 | |
46°C (115°F) | |
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate | |
VXUYXOFXAQZZMF-UHFFFAOYSA-N | |
propan-2-olate;titanium(4+) | |
11026 | |
≥97% | |
Moisture sensitive | |
Titanium (IV) isopropoxide |
16°C to 20°C | |
Colorless to Yellow | |
C12H28O4Ti | |
MFCD00008871 | |
3679474 | |
14,9480 | |
Soluble in anhydrous ethanol, ether, benzene and chloroform; Decomposes in water | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4] | |
284.219 | |
284.23 | |
1.464 | |
0.955 g/mL |
Safety and Handling
GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P370+P378q-P501c
H226-H319-H331-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
missing translation for 'einecsNumber' : 208-909-6
missing translation for 'rtecsNumber' : NT8060000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Titanium(IV) isopropoxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Tetraisopropyl titanate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.