missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) isopropoxide, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11388687
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of Sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.219
MDL Number MFCD00008871
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

Melting Point 16°C to 20°C
Color Colorless
Density 0.955 g/mL
Boiling Point 232°C
Flash Point 46°C (115°F)
Linear Formula Ti[OCH(CH3)2]4
Refractive Index 1.464
Quantity 5 g
UN Number UN2413
Beilstein 3679474
Sensitivity Moisture sensitive
Merck Index 14,9480
Solubility Information Miscible with anhydrous ethanol,chloroform,ether and benzene.
Formula Weight 284.23
Percent Purity 99.995%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Titanium(IV) isopropoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) isopropoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetraisopropyl titanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'