missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) methoxide, 95%

Product Code. 11354899
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11354899

Brand: Thermo Scientific Alfa Aesar 077137.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 992-92-7 | Ti(OCH3)4·(CH3OH)x, x<1

Titanium(IV) methoxide is used for the preparation of polyoxotitanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-92-7
Molecular Formula Ti(OCH3)4·(CH3OH)x, x<1
MDL Number MFCD00014883
Synonym tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide

Specifications

Boiling Point >243°C (52mmHg)
Melting Point 200°C to 210°C
Physical Form Granular/Lump
Quantity 500 g
UN Number UN3181
Flash Point 48°C (118°F)
Solubility Information Slightly soluble in organic solvents.
Percent Purity 95%
Sensitivity Moisture sensitive
Chemical Name or Material Titanium (IV) methoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.