missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) methylphenoxide, tech., Thermo Scientific™

Product Code. p-7080508
Click to view available options
Quantity:
10 g
250 g
1 kg
Unit Size:
10g
1kg
250g
This item is not returnable. View return policy

Product Code. 11423780

Brand: Thermo Scientific Alfa Aesar 089454.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28503-70-0
Molecular Formula C28H28O4Ti
MDL Number MFCD00061534
Synonym Titanium(IV) cresylate

Specifications

Boiling Point 230°C to 240°C (0.01mmHg)
Melting Point -36°C
Color Red
Physical Form Liquid
Quantity 10 g
Flash Point 66°C (150°F)
Formula Weight 476.43
Grade Technical
Sensitivity Moisture sensitive
Density 1.055 g/mL
Chemical Name or Material Titanium (IV) methylphenoxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.