missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) n-butoxide, 98+%, Thermo Scientific Chemicals

Artikelnummer. 10216470
Click to view available options
Quantity:
500 g
1 kg
2 kg
10 kg
Packaging:
Glass bottle
Plastic Drum
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 5593-70-4
Molecular Formula C16H36O4Ti
Molecular Weight (g/mol) 340.35
MDL Number MFCD00009433
InChI Key FPCJKVGGYOAWIZ-UHFFFAOYSA-N
Synonym butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
PubChem CID 25203927
IUPAC Name butan-1-ol;titanium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Spezifikation

Melting Point -75°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 310.0°C to 314.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula Ti[O(CH2)3CH3]4
Refractive Index 1.486
Quantity 500 g
Specific Gravity 0.99
Solubility Information Solubility in water: rapidly hydrolized by water. Other solubilities: miscible with most common organic solvents
Viscosity 50-80 mPa.s (20°C)
Formula Weight 340.35
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Titanium(IV) n-butoxide, 99%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Titanium(IV) n-butoxide
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Butyl titanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'