missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) n-butoxide, 98+%, Thermo Scientific Chemicals

Product Code. 10782261
Click to view available options
Quantity:
500 g
1 kg
2 kg
10 kg
Packaging:
Glass bottle
Plastic Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5593-70-4
Molecular Formula C16H36O4Ti
Molecular Weight (g/mol) 340.35
MDL Number MFCD00009433
InChI Key FPCJKVGGYOAWIZ-UHFFFAOYSA-N
Synonym butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
PubChem CID 25203927
IUPAC Name butan-1-ol;titanium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Specifications

Melting Point -75°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 310.0°C to 314.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula Ti[O(CH2)3CH3]4
Refractive Index 1.486
Quantity 2 kg
Specific Gravity 0.99
Solubility Information Solubility in water: rapidly hydrolized by water. Other solubilities: miscible with most common organic solvents
Viscosity 50-80 mPa.s (20°C)
Formula Weight 340.35
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Titanium(IV) n-butoxide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'