missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) n-butoxide, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
Description
Titanium(IV) n-butoxide is used in the preparation of nanosized titania powders in the anatase form and ferroelectric bismuth titanate thin films. It is also used as intermediates, paint additives, coating additives, processing aids and as process regulators. It is involved in the preparation of nanocrystalline titanium dioxide powders at room temperature.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 310°C to 314°C |
Melting Point | -55°C |
Color | Yellow |
Physical Form | Liquid |
Quantity | 10 g |
Linear Formula | Ti[O(CH2)3CH3]4 |
UN Number | UN2924 |
Beilstein | 4148236 |
Flash Point | 41°C (170°F) |
Solubility Information | Miscible with aliphatic,aromatic,chlorinated and oxygenated solvents. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'