Learn More
Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion, Thermo Scientific Chemicals
Beschreibung
UV protection in clear coatings. Photo electrodes of varying properties can be made using colloidal suspensions of titanium(IV) oxide nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation
Spezifikation
Boiling Point | 368°C |
Quantity | 5 kg |
Solubility Information | Insoluble in water. |
Formula Weight | 79.9 |
Concentration or Composition (by Analyte or Components) | 20 to 35% in H2O |
Density | 1.26 g/mL |
Chemical Name or Material | Titanium (IV) oxide, Colloidal dispersion |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q15I001 | Certificate of Analysis | 30/08/2022 | 044517.A1,044517.36,044517.A7 |
Y01D016 | Certificate of Analysis | 08/12/2021 | 044517.A1,044517.36,044517.A7 |
X17E053 | Certificate of Analysis | 07/12/2021 | 044517.A1,044517.36,044517.A7 |
M26D026 | Certificate of Analysis | 07/12/2021 | 044517.A1,044517.36,044517.A7 |
I26X019 | Certificate of Analysis | 07/12/2021 | 044517.A1,044517.36,044517.A7 |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.