missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals
£78.60 - £639.00
Chemical Identifiers
CAS | 13463-67-7 |
---|---|
Molecular Formula | O2Ti |
Molecular Weight (g/mol) | 79.87 |
MDL Number | MFCD00011269 MFCD00210650 |
InChI Key | GWEVSGVZZGPLCZ-UHFFFAOYSA-N |
Synonym | titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance |
PubChem CID | 26042 |
ChEBI | CHEBI:32234 |
IUPAC Name | dioxotitanium |
SMILES | O=[Ti]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10443355
|
Thermo Scientific Acros
384290500 |
50 g | Plastic bottle |
£78.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173793
|
Thermo Scientific Acros
384292500 |
250 g | Plastic bottle |
£245.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646903
|
Thermo Scientific Acros
384290010 |
1 kg | Plastic drum |
£639.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
13463-67-7 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269 MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |
Specifications
2900.0°C | |
13463-67-7 | |
3.5 to 4.5 (4% dispersion) | |
O2Ti | |
MFCD00011269 MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.88 | |
1.5% max. (105°C, 2 hrs) typically | |
4.23 | |
Titanium(IV) oxide |
1855.0°C | |
White to Yellow | |
Powder | |
TiO2 | |
15, 9628 | |
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
≥99.5% (based on ignited material) (typically) | |
2.0% max. (1000°C, 2 hrs) (based on dried material) typically | |
4.2300g/mL |
Safety and Handling
missing translation for 'einecsNumber' : 236-675-5
RUO – Research Use Only