missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
250 g
Description
Employed as a cross linking agent for cellulose fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 184°C (decomposition) |
Physical Form | Powder |
Quantity | 10 g |
Solubility Information | Soluble in water (6.6g/L (20°C)). |
Formula Weight | 262.12 |
Chemical Name or Material | Titanium (IV) oxide bis (2,4-pentanedionate) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'