missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) oxide, catalyst support, Thermo Scientific Chemicals
Anatase, Typical properties = S.A. > 150m2/g, median pore diameter 140Å, total pore volume 0.38cc/g, packing density 0.90kg/L, contains 1% S (as SO4)
£52.56 - £536.67
Chemical Identifiers
CAS | 1317-70-0 |
---|---|
Molecular Formula | O2Ti |
Molecular Weight (g/mol) | 79.87 |
MDL Number | MFCD00011269,MFCD00210650 |
InChI Key | GWEVSGVZZGPLCZ-UHFFFAOYSA-N |
Synonym | titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance |
PubChem CID | 26042 |
ChEBI | CHEBI:32234 |
IUPAC Name | dioxotitanium |
SMILES | O=[Ti]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450720
|
Thermo Scientific Alfa Aesar
044429.30 |
250 g |
£52.56
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460720
|
Thermo Scientific Alfa Aesar
044429.A1 |
1 kg |
£145.73
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470720
|
Thermo Scientific Alfa Aesar
044429.A7 |
5 kg |
£536.67
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-70-0 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269,MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |
Specifications
1317-70-0 | |
O2Ti | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
Titanium(IV) oxide, 1/8 in. Pellets |
Pellets | |
MFCD00011269,MFCD00210650 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.9 |
Safety and Handling
missing translation for 'einecsNumber' : 215-280-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only