missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) oxide, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
250 g
1 kg
25 kg
6 x 1 kg
Unit Size:
1kg
250g
25kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | >2,000°C |
| Melting Point | >350°C (literature) |
| CAS Max % | 100.0000% |
| Quantity | 25 kg |
| Assay Percent Range | 99 to 100.5% |
| Linear Formula | TiO2 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 79.87 |
| Percent Purity | 99.0 to 100.5% (calc. to the dried substance) |
| Grade | puriss. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction