missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) oxide, rutile, 99.5% min (metals basis), Thermo Scientific Chemicals Product Code.: 11334698

Titanium(IV) oxide, rutile, 99.5% min (metals basis), Thermo Scientific Chemicals

Product Code. 11334698
250 g
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11334698

Brand: Thermo Scientific Alfa Aesar 043047.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Titanium dioxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-80-2
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 2500°C to 3000°C
Melting Point 1830°C to 1850°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5% min (metals basis)
Solubility Information Soluble in hot concentrated sulfuric acid,in hydrofluoric acid and alkali. Insoluble in water.
Formula Weight 79.9
Density 4.23
Chemical Name or Material Titanium(IV) oxide, rutile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ12K005 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number043047.A1,043047.30,043047.A7
Lot NumberX16K033 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number043047.A1,043047.30,043047.A7
Lot NumberE17Z045 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043047.A1,043047.30,043047.A7
Lot NumberP15I035 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043047.A1,043047.30,043047.A7
Lot NumberA23Y028 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043047.A1,043047.30,043047.A7
compliance-icons
Product Identifier
  • Titanium(IV) oxide, rutile
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Rutile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) oxide, rutile, 99.5% min (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.