missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, rutile, 99.8% (metals basis)

Product Code. 11365417
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11365417

Brand: Thermo Scientific Alfa Aesar 042681.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-80-2 | O2Ti | 79.87 g/mol

Titanium(IV) oxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-80-2
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 2500°C to 3000°C
Melting Point 1830°C to 1850°C
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in hot concentrated sulfuric acid,in hydrofluoric acid and alkali. Insoluble in water.
Formula Weight 79.9
Density 4.23 g/mL
Chemical Name or Material Titanium(IV) oxide
What is the difference between Titanium (IV) oxide, rutile and Titanium (IV) oxide, anatase?

Rutile and anatase are polymorphs of TiO2, which are both naturally occurring. Rutile is a source for titanium smelting and is used in the semiconductor industry to produce dye-based solar cells (Grätzel cells). Anatase shows a high photocatalytic activity, which limits the usability as a pigment in polymers, but also opens certain applications as a catalyst. Anatase has a higher photocatalytic activity than rutile but is thermodynamically less stable.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.