Learn More
Titanium(IV) oxide, rutile, 99.9% (metals basis), Thermo Scientific Chemicals
£70.20 - £176.00
Chemical Identifiers
CAS | 1317-80-2 |
---|---|
Molecular Formula | O2Ti |
Molecular Weight (g/mol) | 79.87 |
MDL Number | MFCD00011269,MFCD00210650 |
InChI Key | GWEVSGVZZGPLCZ-UHFFFAOYSA-N |
Synonym | titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance |
PubChem CID | 26042 |
ChEBI | CHEBI:32234 |
IUPAC Name | dioxotitanium |
SMILES | O=[Ti]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458550
|
Thermo Scientific Alfa Aesar
044375.14 |
25 g |
£70.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448550
|
Thermo Scientific Alfa Aesar
044375.22 |
100 g |
£176.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium dioxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-80-2 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269,MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |
Specifications
2500°C to 3000°C | |
1317-80-2 | |
99.9% (metals basis) | |
MFCD00011269,MFCD00210650 | |
Soluble in hot concentrated sulfuric acid,in hydrofluoric acid and alkali. Insoluble in water. | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
4.23 g/mL |
1830°C to 1850°C | |
Sintered Pieces | |
O2Ti | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.9 | |
Titanium(IV) oxide |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'einecsNumber' : 215-282-2
missing translation for 'rtecsNumber' : XR2275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only