missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) tert-butoxide, 98%

Product Code. 10441402 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10441402

Brand: Thermo Scientific Acros 344000050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3087-39-6 | C16H36O4Ti | 340.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3087-39-6
Molecular Formula C16H36O4Ti
Molecular Weight (g/mol) 340.33
MDL Number MFCD00040554
InChI Key GRWPYGBKJYICOO-UHFFFAOYSA-N
Synonym unii-px6yr1s99s, px6yr1s99s, titanium iv tert-butoxide, titanium tetra-tert-butoxide, titanium tert-butoxide, titanum tetra t-butoxide, tetra-tert-butoxytitanium iv, titanium 4+ 2-methylpropan-2-olate
PubChem CID 6451515
SMILES [Ti+4].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]

Specifications

Color Colorless
Density 0.8893g/mL
Boiling Point 160.0°C to 165.0°C (15.0 mmHg)
Flash Point 47°C
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula Ti[OC(CH3)3]4
Quantity 5 g
Beilstein 01, IV, 1613
Specific Gravity 0.8893
IUPAC Name titanium(4+) tetrakis(2-methylpropan-2-olate)
Formula Weight 340.33
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Titanium(IV) tert-butoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(IV)tert-butoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.