Learn More
Titanium nitride, 99.5% (metals basis), Thermo Scientific Chemicals
£63.10 - £1009.00
Chemical Identifiers
CAS | 25583-20-4 |
---|---|
Molecular Formula | NTi |
Molecular Weight (g/mol) | 61.87 |
MDL Number | MFCD00049596 |
InChI Key | NRTOMJZYCJJWKI-UHFFFAOYSA-N |
Synonym | titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a |
PubChem CID | 93091 |
IUPAC Name | azanylidynetitanium |
SMILES | N#[Ti] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344318
|
Thermo Scientific Alfa Aesar
041556.18 |
50 g |
£63.10
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354318
|
Thermo Scientific Alfa Aesar
041556.30 |
250 g |
£263.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364318
|
Thermo Scientific Alfa Aesar
041556.A1 |
1 kg |
£1009.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium nitride is used as a processing agent in powder metallurgy and laboratory. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is used as Use as primary and secondary intermediate. Artificial limbs; Barrier layer in contact and interconnect metallization; Biological materials cutting tools; Gate electrode in metal-oxide-semiconductor (MOS) transistors; Low-barrier Schottky diode; Optical devices in aggressive environments; Plastic molds; Prostheses; Wear-resistant coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25583-20-4 | |
61.87 | |
NRTOMJZYCJJWKI-UHFFFAOYSA-N | |
93091 | |
N#[Ti] |
NTi | |
MFCD00049596 | |
titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a | |
azanylidynetitanium |
Specifications
2950°C | |
Yellow | |
99.5% metal basis | |
MFCD00049596 | |
it is soluble in aqua regia on boiling. | |
N#[Ti] | |
61.87 | |
61.91 | |
Titanium nitride |
25583-20-4 | |
Powder | |
NTi | |
titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a | |
NRTOMJZYCJJWKI-UHFFFAOYSA-N | |
azanylidynetitanium | |
93091 | |
5.22 |
Safety and Handling
missing translation for 'einecsNumber' : 247-117-5
missing translation for 'rtecsNumber' : XR2230000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only