missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Titanium On-line Filter Kit

Used in chromatography of biomolecules. Cytiva Titanium On-line Filter Kit is resistant to most organic solvents and salt buffers commonly found in chromatography applications.

Brand:  Cytiva 18-1120-94

Code : GX

Additional Details : Weight : 0.00090kg

Product Code. 10230165

  • £80.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

  •  Pore size: 2 μm
  • The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
  • Material Titanium (Ti) 
Specifications

Specifications

1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers