missing translation for 'onlineSavingsMsg'
Learn More

Titanium oxide, 99.9% (metals basis)

Product Code. p-7076378
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11349057

Brand: Thermo Scientific Alfa Aesar 039000.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts. | CAS: 12065-65-5 | Ti3O5

Nanocrystalline titanium oxide electrodes is useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12065-65-5
Molecular Formula Ti3O5
MDL Number MFCD00210650
Synonym Trititanium pentoxide

Specifications

Melting Point 1800°C
Physical Form Pieces
Quantity 25 g
Solubility Information Soluble in hot concentrated sulfuric acid,hydrofluoric acid and alkali.
Percent Purity 99.9%
Packaging Plastic bottle
Specific Gravity 4.2
Density 4.2
Chemical Name or Material Titanium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.