missing translation for 'onlineSavingsMsg'
Learn More

Titanium oxide, 99.9% (metals basis)

Product Code. 11398768
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11398768

Brand: Thermo Scientific Alfa Aesar 041635.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For vacuum deposition | CAS: 12065-65-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12065-65-5
MDL Number MFCD00210650
Synonym Trititanium pentoxide

Specifications

Melting Point 1800°C
Color Black
Physical Form Pellets
Quantity 10 g
Linear Formula Ti3O5
Percent Purity 99.9%
Density 4.20 g/mL
Assay (metals basis)
Chemical Name or Material Titanium oxide, Pellets, ≈8 to 9mm dia. x 7 to 9mm thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.