missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium powder, -100 mesh, 99.4%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
UN Number | UN2546 |
Quantity | 25 g |
Percent Purity | 99.4% |
Mesh Size | −100 Mesh |
Odor | Odorless |
Form | Powder |
Chemical Name or Material | Titanium powder |
Safety and Handling

Product Identifier
- Titanium powder
- Danger
- Pyrophoric solid Category 1
- Self-heating substance Category 2
- H250-Catches fire spontaneously if exposed to air.
- H252-Self-heating in large quantities; may catch fire.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use to extinguish.
- P422-Store contents under .
- P420-Store away from other materials.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Titanium powder, -100 mesh, 99.4%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction