missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium powder, <20 micron, 93%, packaged in water, Thermo Scientific Chemicals
£35.68 - £1191.93
Chemical Identifiers
CAS | 7440-32-6 |
---|---|
Molecular Formula | Ti |
Molecular Weight (g/mol) | 47.87 |
MDL Number | MFCD00011264,MFCD00151301 |
InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
PubChem CID | 23963 |
ChEBI | CHEBI:33341 |
IUPAC Name | titanium |
SMILES | [Ti] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306627
|
Thermo Scientific Alfa Aesar
000681.06 |
5 g |
£35.68
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386617
|
Thermo Scientific Alfa Aesar
000681.14 |
25 g |
£89.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396617
|
Thermo Scientific Alfa Aesar
000681.22 |
100 g |
£273.36
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316627
|
Thermo Scientific Alfa Aesar
000681.36 |
500 g |
£1191.93
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-32-6 | |
47.87 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
23963 | |
titanium |
Ti | |
MFCD00011264,MFCD00151301 | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
CHEBI:33341 | |
[Ti] |
Specifications
Gray | |
Dark Gray powder | |
Ti | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
[Ti] | |
47.87 | |
CHEBI:33341 | |
Powder |
7440-32-6 | |
93% | |
MFCD00011264,MFCD00151301 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
titanium | |
23963 | |
Odorless | |
Titanium |
Safety and Handling
P210-P222-P280-P335+P334-P370+P378q
H250-H500
missing translation for 'dotInformation' : DOT Class: 4.2; Packaging Group: II
missing translation for 'einecsNumber' : 231-142-3
missing translation for 'rtecsNumber' : XR1700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Titanium powder
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P370+P378-In case of fire: Use to extinguish.
Supplemental information
- MIXTURE LIST-Contains : Titanium
RUO – Research Use Only