missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, spherical, -150 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11364298
Click to view available options
Quantity:
25 g
100 g
500 g

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

UN Number UN2546
Quantity 100 g
Percent Purity 99.9%
Mesh Size −150 Mesh
Odor Odorless
Form Spherical Powder
Assay (metals basis)
Chemical Name or Material Titanium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'