missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium silicide, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
It is used as source, gate and drain contacts and local interconnections in CMOS integrated circuits. it is used in the C54 phase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 2130°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.5%, metal basis |
Solubility Information | Soluble in hydrofluoric acid. Insoluble in water. |
Formula Weight | 323.76 |
Chemical Name or Material | Titanium Silicide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'