missing translation for 'onlineSavingsMsg'
Learn More
Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length, 99.9+% (metals basis), Thermo Scientific Chemicals Product Code.: 11345838

Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length, 99.9+% (metals basis), Thermo Scientific Chemicals

Product Code. 11345838
1 Slug
Click to view available options
Quantity:
1 Slug
25 Slugs
5 Slugs
This item is not returnable. View return policy

Product Code. 11345838

Brand: Thermo Scientific Alfa Aesar 010381.QI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 1 Slug
Percent Purity ≥99.9%
Odor Odorless
Weight ≈3g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW23I019 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog Number010381.QI,010378.22,010378.36
Lot NumberM17I042 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010381.QI,010378.22,010378.36
Lot NumberX22G036 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010381.QI,010378.22,010378.36
Lot NumberC15R008 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010381.QI,010378.22,010378.36
Lot NumberW29D032 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010381.QI,010378.22,010378.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length, 99.9+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.