Learn More
Titanium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis), Thermo Scientific Chemicals
Description
Titanium sputtering target and its alloys are characterized by their lightness, strength and corrosion resistance and are used widely in aerospace applications. In addition, these properties also make the material suitable for medical applications (e.g. replacement hip joints). Titanium dioxide is used as a white pigment in paints and plastics as it provides great opacity. The same material is also used in the manufacture of heat resisting and durable glass, the TiO2 replacing certain proportions of the soda. Titanium carbide is used to manufacture cemented carbides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 Ea. |
CAS | 7440-32-6 |
Odor | Odorless |
Molecular Formula | Ti |
MDL Number | MFCD00011264,MFCD00151301 |
Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
Solubility Information | Insoluble in water. |
InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
SMILES | [Ti] |
IUPAC Name | titanium |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
C07R027 | Certificate of Analysis | 28/12/2021 | 043042.KS |
C04X024 | Certificate of Analysis | 28/12/2021 | 043042.KS |
H30K17 | Certificate of Analysis | 08/12/2021 | 043042.KS |
D12X042 | Certificate of Analysis | 08/12/2021 | 043042.KS |
G23Z030 | Certificate of Analysis | 07/12/2021 | 043042.KS |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.