missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Ea.
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Especificaciones
Especificaciones
Quantity | 1 Ea. |
CAS | 7440-32-6 |
Odor | Odorless |
Molecular Formula | Ti |
MDL Number | MFCD00011264,MFCD00151301 |
Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
SMILES | [Ti] |
IUPAC Name | titanium |
Molecular Weight (g/mol) | 47.87 |
Mostrar más |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
B24P24 | Certificate of Analysis | 18/11/2021 | 043044.KS |
X25E018 | Certificate of Analysis | 18/11/2021 | 043044.KS |
E05T003 | Certificate of Analysis | 18/11/2021 | 043044.KS |
L10T017 | Certificate of Analysis | 18/11/2021 | 043044.KS |
E05R025 | Certificate of Analysis | 18/11/2021 | 043044.KS |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Titanium sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis), Thermo Scientific Chemicals >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido